Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SDPR Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA584530
Description
Immunogen sequence: QHPGSDMRQE KPSSPSPMPS STPSPSLNLG NTEEAIRDNS QVNAVTVLTL LDKLVNMLDA VQENQHKMEQ RQISLEGSVK GIQNDLT.
The serum deprivation-response protein (SDPR) is a calcium-independent phospholipid-binding protein whose expression is increased in serum-starved cells. SDPR is a substrate for protein kinase C (PKC) phosphorylation and recruits the polymerase I and transcript release factor (PTRF) to caveolae. Removal of this protein causes caveolae loss and its over-expression results in caveolae deformation and membrane tubulation. Both SDPR and PTRF, as well as the other member of the cavin family PRKCDBP were down regulated in breast cancer cell lines and breast tumor tissue, suggesting that expression of the cavin family proteins could be a useful prognostic indicator of breast cancer progression.
Specifications
SDPR | |
Polyclonal | |
Unconjugated | |
CAVIN2 | |
caveolae associated protein 2; caveolae-associated protein 2; cavin 2; CAVIN2; cavin-2; EMBL:AAH81956.1}; phosphatidylserine binding protein; phosphatidylserine-binding protein; PS-p68; SDPR; sdpr {ECO:0000312; SDR; serum deprivation response; serum deprivation response protein; serum deprivation-response protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8436 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O95810 | |
CAVIN2 | |
Recombinant protein corresponding to Human CAVIN2. Recombinant protein control fragment (Product #RP-107133). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction