Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SDPR Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984351
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4351 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4351 Supplier Bioss Supplier No. BS19605R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The serum deprivation-response protein (SDPR) is a calcium-independent phospholipid-binding protein whose expression is increased in serum-starved cells. SDPR is a substrate for protein kinase C (PKC) phosphorylation and recruits the polymerase I and transcript release factor (PTRF) to caveolae. Removal of this protein causes caveolae loss and its over-expression results in caveolae deformation and membrane tubulation. Both SDPR and PTRF, as well as the other member of the cavin family PRKCDBP were down regulated in breast cancer cell lines and breast tumor tissue, suggesting that expression of the cavin family proteins could be a useful prognostic indicator of breast cancer progression.

Specifications

Antigen SDPR
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CAVIN2
Gene Accession No. O95810
Gene Alias BOS_1929; caveolae associated protein 2; Caveolae-associated protein 2; cavin 2; CAVIN2; Cavin-2; EMBL:AAH81956.1}; phosphatidylserine binding protein; Phosphatidylserine-binding protein; PS-p68; Sdpr; sdpr {ECO:0000312; Sdr; serum deprivation response; serum deprivation response protein; Serum deprivation-response protein
Gene Symbols CAVIN2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SDPR.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8436
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.