Learn More
Invitrogen™ SDR Polyclonal Antibody

Description
SDR Polyclonal Antibody for Western Blot, ELISA
Specifications
Specifications
| Antigen | SDR |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 4.04 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 |
| Gene Alias | Short-chain alcohol dehydrogenase; Short-chain dehydrogenase |
| Host Species | Rabbit |
| Immunogen | Recombinant Zebrafish Short-chain alcohol dehydrogenase protein (1-137AA). |
| Purification Method | Protein G |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.