Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SDR16C5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555229
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555229 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555229 Supplier Invitrogen™ Supplier No. PA555229
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CTTGCPSLLP ILEPKYAVEK IVEAILQEKM YLYMPKLLYF MMFLKSFLPL KTGLLIADYL GILHAMDGFV DQKKKL Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 76%.

RDHE2 belongs to a family of short-chain alcohol dehydrogenases/reductases that catalyze the first and rate-limiting step that generates retinaldehyde from retinol (Matsuzaka et al., 2002 [PubMed 12372410]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SDR16C5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SDR16C5
Gene Accession No. Q8N3Y7
Gene Alias EPHD-2; epidermal retinal dehydrogenase 2; epidermal retinol dehydrogenase 2; RDH#2; RDHE2; RDH-E2; retinal short chain dehydrogenase reductase; retinal short chain dehydrogenase reductase (Rdhe2); retinal short chain dehydrogenase reductase 2; retinal short-chain dehydrogenase reductase 2; retSDR2; Scdr9; Sdr16c5; short chain dehydrogenase reductase 9; short chain dehydrogenase/reductase family 16C, member 5; short-chain dehydrogenase reductase 9; Short-chain dehydrogenase/reductase family 16C member 5
Gene Symbols SDR16C5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SDR16C5. Recombinant protein control fragment (Product #RP-93897).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 195814
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.