Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SDR42E2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563941
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563941 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563941 Supplier Invitrogen™ Supplier No. PA563941
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VASYGMSGAE KLQKEQIESI NVGGTKLVID VCVR Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 53%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen SDR42E2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SDR42E2
Gene Accession No. A6NKP2
Gene Alias Putative 3-beta-HSD family protein ENSP00000330812; putative short-chain dehydrogenase/reductase family 42E member 2; SDR42E2; short chain dehydrogenase/reductase family 42E, member 2
Gene Symbols SDR42E2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SDR42E2. Recombinant protein control fragment (Product #RP-101845).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100288072
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.