Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SDR9C7 Antibody (OTI4B5), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274036V
Description
SDR9C7 Monoclonal specifically detects SDR9C7 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.Specifications
SDR9C7 | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SDR9C7 | |
Full length recombinant protein of human SDR9C7 (NP_683695) produced in E.coli. | |
0.1 mL | |
121214.0 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, Immunohistochemistry | |
OTI4B5 | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
EC 1.1.1, EC 1.1.1.-, FLJ16333, MGC126600, Orphan short-chain dehydrogenase/reductase, RDH-S, RDHSMGC126602, SDR-Oorphan short-chain dehydrogenase/ reductase, SDROretinol dehydrogenase similar protein, short chain dehydrogenase/reductase family 9C, member 7, short-chain dehydrogenase/reductase family 9C member 7 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction