Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SDR9C7 Monoclonal Antibody (OTI10D2), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA501690S


Description
SDR9C7 Monoclonal antibody specifically detects SDR9C7 in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
SDR9C7 | |
Monoclonal | |
0.85 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
O60884 | |
Mouse | |
Affinity chromatography | |
RUO | |
10294 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Flow Cytometry, Western Blot | |
OTI10D2 | |
Unconjugated | |
DNAJA2 | |
1810054F20Rik; 4short chain dehydrogenase/reductase family 9C, member 7; orphan short chain dehydrogenase/reductase; orphan short-chain dehydrogenase / reductase; orphan short-chain dehydrogenase/reductase; Rdh20; Rdhs; RDH-S; retinol dehydrogenase 20; retinol dehydrogenase similar protein; Sdr9c7; SDRO; SDR-O; short chain dehydrogenase/reductase family 9C member 7; short chain dehydrogenase/reductase family 9C, member 7; short-chain dehydrogenase/reductase family 9C member 7 | |
Full length human recombinant protein of human DNAJA2 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction