Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SDR9C7 Monoclonal Antibody (OTI10F11), TrueMAB™, OriGene
SDP

Catalog No. 501673103
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-3103 100 μL
50-254-1858 30 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-167-3103 Supplier OriGene Supplier No. TA502015
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SDR9C7 encodes a protein with similarity to the short-chain dehydrogenase/reductase (SDR) family but has not been shown to have retinoid or dehydrogenase activities.

Specifications

Antigen SDR9C7
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI10F11
Concentration 0.76 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene SDR9C7
Gene Accession No. Q8NEX9
Gene Alias ARCI13, RDHS, SDR-O, SDRO
Gene Symbols SDR9C7
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 1-313 of human SDR9C7 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 121214
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.