Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SDR9C7 Monoclonal Antibody (OTI4H9), TrueMAB™, OriGene
SDP

Catalog No. 502541859
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-1859 30 μL
50-167-3102 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-1859 Supplier OriGene Supplier No. TA502014S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SDR9C7 Monoclonal antibody specifically detects SDR9C7 in Human samples. It is validated for Western Blot

Specifications

Antigen SDR9C7
Applications Western Blot
Classification Monoclonal
Clone OTI4H9
Concentration 0.79 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene SDR9C7
Gene Accession No. Q8NEX9
Gene Alias 1810054F20Rik; 4short chain dehydrogenase/reductase family 9C, member 7; orphan short chain dehydrogenase/reductase; orphan short-chain dehydrogenase / reductase; orphan short-chain dehydrogenase/reductase; Rdh20; Rdhs; RDH-S; retinol dehydrogenase 20; retinol dehydrogenase similar protein; Sdr9c7; SDRO; SDR-O; short chain dehydrogenase/reductase family 9C member 7; short chain dehydrogenase/reductase family 9C, member 7; short-chain dehydrogenase/reductase family 9C member 7
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-313 of human produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 121214
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.