Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SDR9C7 Monoclonal Antibody (OTI7E11), TrueMAB™, OriGene
SDP

Catalog No. 502542061
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-2061 30 μL
50-167-2850 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-2061 Supplier OriGene Supplier No. TA501732S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SDR9C7 Monoclonal antibody specifically detects SDR9C7 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot

Specifications

Antigen SDR9C7
Applications Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI7E11
Concentration 0.92 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene GGPS1
Gene Accession No. O95749
Gene Alias 1810054F20Rik; 4short chain dehydrogenase/reductase family 9C, member 7; orphan short chain dehydrogenase/reductase; orphan short-chain dehydrogenase / reductase; orphan short-chain dehydrogenase/reductase; Rdh20; Rdhs; RDH-S; retinol dehydrogenase 20; retinol dehydrogenase similar protein; Sdr9c7; SDRO; SDR-O; short chain dehydrogenase/reductase family 9C member 7; short chain dehydrogenase/reductase family 9C, member 7; short-chain dehydrogenase/reductase family 9C member 7
Host Species Mouse
Immunogen Full length human recombinant protein of human GGPS1 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9453
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.