Learn More
SDR9C7 Mouse anti-Human, Clone: OTI11H10, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF501734

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
SDR9C7 encodes a protein with similarity to the short-chain dehydrogenase/reductase (SDR) family but has not been shown to have retinoid or dehydrogenase activities.Specifications
SDR9C7 | |
Monoclonal | |
Unconjugated | |
SDR9C7 | |
ARCI13, RDHS, SDR-O, SDRO | |
Mouse | |
Affinity Chromatography | |
RUO | |
121214 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Western Blot | |
OTI11H10 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q8NEX9 | |
SDR9C7 | |
Full length human recombit protein of human SDR9C7 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.