Learn More
ZING™ Enterprises SDS Binder
Description
ZING SDS Binder prominently stores important safety and training documents to comply with the new GHS/Hazard Communication requirements. Binder is constructed from high quality 100% post-consumer poly materials that reinforce sustainability initiatives as well as compliance. Binder is highly visible with its bright red/white colors and is extremely durable, chemical resistant and easy to clean.
Different sizes include – 1.5 in. (up to 300 sheets), 2.5 in. (up to 500 sheets), and 3 in. (up to 600 sheets). Binders are available in English and bilingual English/Spanish versions.
Specifications
Specifications
| Description | Floor standing |
| Product Type | Hazard Communication |
| Diameter (English) Ring | 1.5 in. |
| Width (English) | 1.5 in. |
| Height (English) | 12 in. |
| Quantity | 1 |
| Material | Plastic |
| Color | Red, White |
| Languages | English |
| For Use With (Application) | SDS Management |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.