Learn More
Invitrogen™ SDS Monoclonal Antibody (OTI2C11)

Description
SDS Monoclonal Antibody for Flow, ICC/IF, IHC (P), Western Blot
Specifications
Specifications
| Antigen | SDS |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI2C11 |
| Concentration | 0.56 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, 1% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | SDS |
| Gene Accession No. | P20132 |
| Gene Alias | 4432411H13Rik; L-serine ammonia-lyase; L-serine deaminase; L-serine dehydratase; L-serine dehydratase/L-threonine deaminase; L-threonine dehydratase; RATSDHE1; Sdh; SDH2; Sdhe1; SDS; serine dehydratase; TDH |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.