Learn More
Cytiva SDS and Native PAGE, IEF kit
Catalog No. 45000447 Shop All Cytiva Products
Change view
Click to view available options
For Use With (Equipment):
Multiphor II
Catalog No. For Use With (Equipment)
45-000-447 Multiphor II
1 options
Catalog No. 45-000-447 Supplier Cytiva Supplier No. 18110245
Includes
  • 2 x Glass plate (125 x 260 x 0.5mm, U-frame)
  • 15 x Glass plate (125 x 260 x 1mm)
  • 2 x Glass plate (125 x 260 x 3mm)
  • 6 x FlexiClamp
  • 100 x IEF electrode strip
  • 500 x EPH paper electrode wick (104 x 253mm)
  • 200 x Sample application piece
  • 5 x IEF/SDS sample application strip (52 samples, 5-20 μl)
  • 50 x Cellophane sheet (210 x 320mm)
Cast homogeneous or gradient polyacrylamide gels (260 x125 x0.5 mm) for SDS PAGE, native PAGE and IEF
Sample application strips and paper pieces are included as well as cellophane sheets for convenient preserving of the gel. GelBond PAGfilm is an optional accessory that allows casting of gels on a plastic backing.
  • Gels cast with or without sample wells
  • Allows casting of gels on a plastic backing
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) SDS, native page
For Use With (Equipment) Multiphor II
Type SDS and Native PAGE, IEF Casting Kit
Includes 2 glass plate (125 x 260 x 0.5mm, U-frame), 15 glass plate (125 x 260 x 1mm), 2 glass plate (125 x 260 x 3mm), 6 FlexiClamp, 100 IEF electrode strip, 500 EPH paper electrode wick (104 x 253mm), 200 sample application piece, 5 IEF/SDS sample application strip (52 samples, 5 to 20μL), 50 cellophane sheet (210 x 320mm), gels cast with or without sample well
Quantity 1 Kit