Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SDS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558704
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558704 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558704 Supplier Invitrogen™ Supplier No. PA558704
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PATIVVPSTT PALTIERLKN EGATVKVVGE LLDEAFELAK ALAKNNPGWV YIPPFDDPLI WEGHASIVKE LKE Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 89%.

SDS encodes one of three enzymes that are involved in metabolizing serine and glycine. L-serine dehydratase converts L-serine to pyruvate and ammonia and requires pyridoxal phosphate as a cofactor. The encoded protein can also metabolize threonine to NH4+ and 2-ketobutyrate. The encoded protein is found predominantly in the liver.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SDS
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SDS
Gene Accession No. P20132
Gene Alias 4432411H13Rik; L-serine ammonia-lyase; L-serine deaminase; L-serine dehydratase; L-serine dehydratase/L-threonine deaminase; L-threonine dehydratase; RATSDHE1; Sdh; SDH2; Sdhe1; SDS; serine dehydratase; TDH
Gene Symbols SDS
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SDS. Recombinant protein control fragment (Product #RP-97731).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10993
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.