Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SDS3 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB100257AF532
Description
SDS3 Polyclonal antibody specifically detects SDS3 in Human, Mouse samples. It is validated for Western BlotSpecifications
SDS3 | |
Polyclonal | |
Western Blot | |
45 kDa Sin3-associated polypeptide, FLJ00052, SAP45suppressor of defective silencing 3 homolog (SDS3, S. cerevisiae), SDS3MGC104711, sin3 histone deacetylase corepressor complex component SDS3, sin3A-associated protein, 45kDa, suppressor of defective silencing 3 homolog (S. cerevisiae), Suppressor of defective silencing 3 protein homolog | |
The immunogen recognized by this antibody maps to a region between residues 175 and 225 of human Suppressor of Defective Silencing 3 using the numbering given in entry NP_071936.2 (GeneID 64426). | |
0.1 mL | |
DNA Repair | |
64426 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Goat | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction