Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SDS3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP24875125UL
Description
SDS3 Polyclonal antibody specifically detects SDS3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
SDS3 | |
Polyclonal | |
Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
45 kDa Sin3-associated polypeptide, FLJ00052, SAP45suppressor of defective silencing 3 homolog (SDS3, S. cerevisiae), SDS3MGC104711, sin3 histone deacetylase corepressor complex component SDS3, sin3A-associated protein, 45kDa, suppressor of defective silencing 3 homolog (S. cerevisiae), Suppressor of defective silencing 3 protein homolog | |
This antibody was developed against a recombinant protein corresponding to amino acids: KAAVKEFEDKKVELKENLIAELEEKKKMIENEKLTMELTGDSMEVKPIMTRKLRRRPNDPVPIPDKRRKPAPAQLNYLLTDEQIMEDLRTLNKLKSPKRPA | |
25 μL | |
DNA Repair | |
64426 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2), 40% Glycerol | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction