Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SDS3 Antibody, Novus Biologicals™

Goat Polyclonal Antibody has been used in 3 publications
Supplier: Novus Biologicals NB100257
Description
SDS3 Polyclonal specifically detects SDS3 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
SDS3 | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
45 kDa Sin3-associated polypeptide, FLJ00052, SAP45suppressor of defective silencing 3 homolog (SDS3, S. cerevisiae), SDS3MGC104711, sin3 histone deacetylase corepressor complex component SDS3, sin3A-associated protein, 45kDa, suppressor of defective silencing 3 homolog (S. cerevisiae), Suppressor of defective silencing 3 protein homolog | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse | |
IgG |
Western Blot, Immunoprecipitation | |
1.0 mg/mL | |
Western Blot 1:1000-1:5000, Immunoprecipitation 2 - 10 ug/mg lysate | |
NP_071936.2 | |
SUDS3 | |
The immunogen recognized by this antibody maps to a region between residues 175 and 225 of human Suppressor of Defective Silencing 3 using the numbering given in entry NP_071936.2 (GeneID 64426). | |
100 μL | |
DNA Repair | |
64426 | |
Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction