Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SDS3 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501559679
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-155-9679 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-155-9679 Supplier Bethyl Laboratories, Inc Supplier No. A301777AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Bovine and Orangutan

SDS3 is a subunit of the histone deacetylase (see HDAC1; MIM 601241)-dependent SIN3A . corepressor complex (Fleischer et al., 2003 [PubMed 12724404]).

Specifications

Antigen SDS3
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene SUDS3
Gene Accession No. Q9H7L9
Gene Alias 45 kDa Sin3-associated polypeptide, FLJ00052, SAP45, SDS3, sin3A-associated protein, 45kDa, suppressor of defective silencing 3 homolog (S. cerevisiae), suppressor of defective silencing 3 homolog (SDS3, S. cerevisiae), suppressor of defective silencing 3 protein homolog
Gene Symbols SUDS3
Host Species Rabbit
Immunogen Between 150 and 200
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64426
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.