Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SDS3 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA572013
Description
SDS3 is a subunit of the histone deacetylase (see HDAC1; MIM 601241)-dependent SIN3A . corepressor complex (Fleischer et al., 2003 [PubMed 12724404]).
Specifications
SDS3 | |
Polyclonal | |
Unconjugated | |
SUDS3 | |
Sin3 histone deacetylase corepressor complex component SDS3; 45 kDa Sin3-associated polypeptide; Suppressor of defective silencing 3 protein homolog; SUDS3; SAP45; SDS3 | |
Rabbit | |
Protein A | |
RUO | |
64426, 71954 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.35 mg/mL | |
PBS with 0.09% sodium azide | |
Q8BR65, Q9H7L9 | |
SUDS3 | |
KLH conjugated synthetic peptide between 72-98 amino acids from the N-terminal region of human SUDS3 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction