Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SDSL Monoclonal Antibody (OTI1A8), TrueMAB™, OriGene
SDP

Catalog No. 502539865
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-253-9865 30 μL
50-167-5713 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-253-9865 Supplier OriGene Supplier No. TA505179S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SDSL Monoclonal antibody specifically detects SDSL in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen SDSL
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1A8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene RIOK2
Gene Accession No. Q9BVS4
Gene Alias 4432411H13Rik; AI504310; cSDH; L-serine deaminase; L-serine dehydratase/L-threonine deaminase; L-threonine dehydratase; SDH; SDH 2; SDH1; Sds; SDSL; SDS-RS1; Serine dehydratase 2; serine dehydratase like; serine dehydratase related sequence 1; serine dehydratase-like; TDH
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 277-552 of human RIOK2 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55781
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.