Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SDSL Monoclonal Antibody (OTI1G4)
GREENER_CHOICE

Catalog No. PIMA526076
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526076 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526076 Supplier Invitrogen™ Supplier No. MA526076
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SDSL Monoclonal Antibody for Western Blot, ICC/IF

SDSL has low serine dehydratase and threonine dehydratase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SDSL
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI1G4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SDSL
Gene Accession No. Q96GA7
Gene Alias 4432411H13Rik; AI504310; cSDH; L-serine deaminase; L-serine dehydratase/L-threonine deaminase; L-threonine dehydratase; SDH; SDH 2; SDH1; Sds; SDSL; SDS-RS1; Serine dehydratase 2; serine dehydratase like; serine dehydratase related sequence 1; serine dehydratase-like; TDH
Gene Symbols SDSL
Host Species Mouse
Immunogen Full length human recombinant protein of SDSL produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 113675
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.