Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SDSL Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542765
Description
Peptide sequence: MDGPVAEHAK QEPFHVVTPL LESWALSQVA GMPVFLKCEN VQPSGSFKIR Sequence homology: Human: 100%.
SDSL has low serine dehydratase and threonine dehydratase activity.
Specifications
SDSL | |
Polyclonal | |
Unconjugated | |
SDSL | |
4432411H13Rik; AI504310; cSDH; L-serine deaminase; L-serine dehydratase/L-threonine deaminase; L-threonine dehydratase; SDH; SDH 2; SDH1; Sds; SDSL; SDS-RS1; Serine dehydratase 2; serine dehydratase like; serine dehydratase related sequence 1; serine dehydratase-like; TDH | |
Rabbit | |
Affinity Chromatography | |
RUO | |
113675 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96GA7 | |
SDSL | |
Synthetic peptide directed towards the N-terminal of human SDSL (aa 1-50). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction