Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SDSL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112706
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112706 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112706 Supplier Invitrogen™ Supplier No. PA5112706
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SDSL Polyclonal Antibody for Western Blot, IHC (P), ELISA

SDSL has low serine dehydratase and threonine dehydratase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SDSL
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene SDSL
Gene Accession No. Q8R238, Q96GA7
Gene Alias 4432411H13Rik; AI504310; cSDH; L-serine deaminase; L-serine dehydratase/L-threonine deaminase; L-threonine dehydratase; SDH; SDH 2; SDH1; Sds; SDSL; SDS-RS1; Serine dehydratase 2; serine dehydratase like; serine dehydratase related sequence 1; serine dehydratase-like; TDH
Gene Symbols SDSL
Host Species Rabbit
Immunogen Recombinant Human Serine dehydratase-like protein (39-169AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 113675, 257635
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.