Learn More
Cytiva SE 250 Mini-Vertical Unit for Two Slab Gels, complete
Catalog No. 45001714 Shop All Cytiva Products
Change view
Click to view available options
SE 250 Mini-Vertical Unit for two slab gels
Catalog No.
45-001-714 SE 250 Mini-Vertical Unit for two slab gels
1 options
Catalog No. 45-001-714 Supplier Cytiva Supplier No. 80614745
Includes
Basic unit, 10 glass plates, two alumina notched plates, well-locating decal, SE 245 Dual Gel Caster, two each 0.75mm thick 10-well combs and 0.75mm thick spacer set.
SE 250 Mini-Vertical unit for two slab gels
  • Premium, multi-option, mini-vertical electrophoresis unit
  • Choose the SE 250 for fast results using a 10 x 8 cm mini-gel format
  • Run two gels at once in a single unit, and rely on the efficient cooling system to deliver flat, side-by-side sample lanes with no smiling
  • Assemble gel sandwiches and electrophoresis units quickly and easily using T-shaped spacers and specially designed clamps with integral springs
  • Cast up to four gels at once using a variety of casting stands or use precast gels from other manufacturers
  • Trust the safety lid interlocks to prevent accidental operator contact
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance The product fulfills valid directives and standards when used within the conditions specified in the user manual. The product must also be used in the same state as it was delivered from Cytiva and connected only to other CE labeled Cytiva m
Running Time 1 to 2 hr.
Type Mini Vertical Electrophoresis Unit
Humidity Range <0.8
Includes Basic unit, 10 glass plates, two alumina notched plates, well-locating decal, SE 245 Dual Gel Caster, two each 0.75-mm thick 10-well combs and 0.75-mm thick spacer set.
Voltage 500 V
Wattage <12 w