Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva SE 400 Vertical Slab Gel Unit

Catalog No. 45001894 Shop All Cytiva Products
Change view
Click to view available options
Voltage:
500 VDC
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Voltage
45-001-894 500 VDC
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-001-894 Supplier Cytiva Supplier No. 80615524

For use in molecular characterisation, purity confirmation and quantitative determination. Cytiva SE 400 Vertical Slab Gel Unit is injection-moulded of tough, impact-resistant plastics to withstand rigours of daily lab use.

  • Sturdier vertical slab gel unit was designed with durability
  • Secure vertical slab electrophoresis units with many of conveniences of SE 600 series units
  • Rugged injection-molded construction for easy assembly and durability in high-use labs
  • Built-in casting stand seals sandwiches leak-free without tape
  • Plates, clamps, spacers and combs are interchangeable with SE 600 series units
  • SE 400 Vertical Units run one 16cm gel or two gels with accessory divider plate
  • Convenient casting stand for single gels
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Fulfills valid directives and standards when used within the conditions specified in the user manual. Product must be used in the same state as it was delivered from Cytiva. Connected only to other CE labeled Cytiva modules or other products as recommended.
For Use With (Application) Molecular characterisation, purity confirmation and quantitative determination
Gel Size 16 cm
Running Time 3 to 5 hr.
Type Slab Gel Vertical Unit, Complete
Includes One Set of Glass Plates (18 x 16 cm), 2 Clamp Assemblies, 2 Cams, 15-Well Comb, and 2 Spacers (1.5 mm)
Model SE400-15-1.5
No. of Gels Up to 2
No. of Wells 15
Quantity 1 Ea.
Temperature (Metric) Operating 0°C to 45°C
Volume (Metric) Lower Buffer Chamber Max. 350 mL
Volume (Metric) Lower Buffer Chamber Min. 150 mL
Volume (Metric) Sample Buffer volume:350 mL (Minimum upper buffer volume)150 mL (Minimum lower buffer volume) 350 mL (Maximum lower buffer volume)
Volume (Metric) Upper Buffer Chamber 350 mL
Weight (English) 7 lb.
Weight (Metric) 3.2 kg
Voltage 500 VDC
Wattage <20 w
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.