Learn More
Cytiva SE260/SE250 MiniVertical Electrophoresis System Replacement Parts: Plates
GSA_VA
Catalog No. 45001887 Shop All Cytiva Products
Change view
Click to view available options
For Use With (Equipment):
HE260 Mini Vertical Unit
SE 250 Mini Vertical Gel Electrophoresis Unit
Type:
Glass Plate
Notched Alumina Plate
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Type
45-001-887 SE 250 Mini Vertical Gel Electrophoresis Unit Glass Plate
45-001-886 HE260 Mini Vertical Unit Notched Alumina Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-001-887 Supplier Cytiva Supplier No. 80615087
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) SE 250 Mini Vertical Gel Electrophoresis Unit
Type Glass Plate
Material Glass
Shape Rectangular
Length (Metric) 10.5 cm
Width (Metric) 10 cm