Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Epredia™ 10% Neutral Buffered Formalin in Seal-Rite Containers
Use these containers for the transportation and short-term storage of patient specimens.
$74.75 - $317.00
Catalog Number | Mfr. No. | Product Type | Packaging Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Packaging Type | Price | Quantity & Availability | ||||
22-110-757
|
Epredia™
51201 |
Formalin | Carton; 192/cs; Cs. dimensions 13-5/16 x 10-1/16 x 7-3/4 in; Cs. weight 10 lbs |
Case of 192 for $172.00
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
22-045-400
|
Epredia™
LC0020 |
Formalin | Tray; 784/cs; Cs. dimensions 25-5/16 x 15-7/16 x 13-1/8 in; Cs. weight 38 lbs |
Case of 784 for $807.00
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
9990910
|
Epredia™
9990910 |
Specimen Storage Containers | Tray; 180/cs; Cs. dimensions 14-13/16 x 14-13/16 x 18-1/4 in; Cs. weight 20 lbs |
Case of 180 for $317.00
|
|
||||
29-016
|
Epredia™
29016 |
Prostate Kit | Tray; 192/cs; Cs. dimensions 12-11/16 x 9-9/16 x 7-1/16 in; Cs. weight: 10 lbs |
Each for $74.75
|
|
||||
Description
Features:- Seal-Rite containers are engineered to maintain seal at 95KPA – the industry standard for proper sealing and transportation via air shipment
- Double-click feature provides audible confirmation that the lid has been aligned, tightened properly and sealed
- Arrow and lock icons align to visually signify the containers have been properly sealed
- Embossed mL measuring scale identifies volume of liquid in the container
- Knurled cap and base of the jar for better gripping
- Safety. Security. Peace of Mind – Avoid the loss of precious tissue samples and exposure of staff to chemicals caused by leaking containers
The containers have been tested using industry-standard International Safe Transit Association (ISTA) performance validation. Internal tests were performed as well with 0% leakage rate.
Specifications
20mL |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title