Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ BRAND™ Sealing films for microplates and PCR plates

Catalog No. p-7146498
Click to view available options
Material:
Polyester
Polyolefin
Vinyl
No. per Pack:
100 Sheets
5 Packs of 10 Sheets
50 4-Strips

Premium sealing films for microplates, deepwell plates and PCR plates

  • Polyester film is optimized for ELISA applications, and is available plain, with colored strips for plate tracking, or in strips for easy well access
  • Vinyl film that is piereceable/resealable for use in automation applications.
  • Polyolefin film is highly transparent and suitable for real time qPCR and protein crystallization. It features a special non-tacky surface for easy placement and reduced chance of sample contaimination.
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Solvent Or Alcohol Resistant DMSO Resistant
Material Polyester
Sterility Non-sterile
Autoclavable Not Autoclavable
No. of Wells 96/384
No. per Pack 100 Sheets
Format ANSI/SLAS
Pre-slit No
Product Type Sealing Film
Adhesive Pressure Sensitive
Certified RNase/DNase-free Yes
Pierceable No
Temperature Range (Metric) -40°C to +120°C
Well Shape All well shapes
Plate Compatibility Deep Well Plate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.