Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BRANDTECH™ BRAND™ Sealing films for microplates and PCR plates
Shop All BrandTech Scientific Products
Click to view available options
Material:
Polyester
Polyolefin
Vinyl
No. per Pack:
100 Sheets
5 Packs of 10 Sheets
50 4-Strips
Description
- Polyester film is optimized for ELISA applications, and is available plain, with colored strips for plate tracking, or in strips for easy well access
- Vinyl film that is piereceable/resealable for use in automation applications.
- Polyolefin film is highly transparent and suitable for real time qPCR and protein crystallization. It features a special non-tacky surface for easy placement and reduced chance of sample contaimination.
Specifications
Specifications
| Color | Clear |
| Solvent Or Alcohol Resistant | No Resistance |
| Material | Vinyl |
| Sterility | Non-sterile |
| Autoclavable | Not Autoclavable |
| No. of Wells | 96 |
| No. per Pack | 100 Sheets |
| Format | ANSI/SLAS |
| Pre-slit | Yes |
| Product Type | Sealing Film |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction