Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Seamless Cellulose Dialysis Tubing
SCI_ED

Catalog No. S25645E Shop All Fisherbrand Products
Click to view available options
Diameter (Metric):
6.4 mm
15.9 mm
20.4 mm
28.6 mm
32 mm
45 mm
47.7 mm
Length (Metric):
3 m
30 m

Reliable and flexible dialysis tubing with 12,000 MWCO for separating macromolecules in educational chemistry, biology, and pharmaceutical labs.

  • Material: Seamless cellulose
  • Molecular Weight Cut-Off (MWCO): Retains molecules ≥12,000
  • Da Applications: Ideal for educational settings in analytical chemistry, biology, and pharmaceutical studies
  • Permeability: High permeability to water and low molecular weight compounds
  • Pore Diameter: Approx. 4.8 nm
  • Formulation: Glycerin, water, and 1% sulfur
  • Storage: Store in an airtight container; rehumidify if dried out
  • Diameter: 45 mm (1.13 in.)
  • Length: 30 m (100 ft.)
  • Quantity: Sold individually (Each)
  • Educational Use Only: No Certificate of Analysis (C of A) provided

This product is for educational use - no C of A is available.

Specifications

Product Type Dialysis Tubing
Material Seamless Cellulose
Diameter (Metric) 6.4 mm
Diameter (English) 0.25 in.
Length (English) 100 ft.
Length (Metric) 30 m
Formulation Glycerin, water and 1% sulfur
Quantity Each
For Use With (Application) Useful in analytical chemistry, biology, and pharmaceutical fields
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.