Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisher Science Education™ Seamless Cellulose Dialysis Tubing PROMO
Shop All Fisherbrand Products

Click to view available options
Length (English):
100 ft.
Description
- Especially useful in analytical chemistry, biology, and pharmaceutical fields
- Highly permeable to water and low molecular weight compounds
- Contains glycerin, water and 1% sulfur
- Must be stored in airtight container or rehumidified before use
- Pore diameter: 4.8 nm
Specifications
Specifications
| Product Type | Dialysis Tubing |
| Material | Cellulose |
| Diameter (Metric) | 15.9 mm |
| Diameter (English) | 0.62 in. |
| Length (English) | 100 ft. |
| Length (Metric) | 30 m |
| Formulation | Glycerin, water and 1% sulfur |
| Quantity | 1 Ea. |
| For Use With (Application) | Analytical chemistry, biology, and pharmaceutical fields |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction