Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Economy Weight Seamless Knit Cotton/Polyester Gloves

Catalog No. 19046441 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-046-441 Small
19-046-442 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-046-441 Supplier PIP™ Supplier No. 35C103/S
Add to Cart
Add to Cart
This item is not returnable. View return policy

Seamless knit shell is comfortable and flexible, while the knit wrist helps prevent dirt and debris from entering the glove.

  • Reversible design allows gloves to be replaced one glove at a time, rather than in pairs
  • Design is thin enough to be worn as a glove liner
  • Use for warehousing, parts handling, assembly, and food processing
  • Complies with FDA food handling requirements 21 CFR, Part 177
  • Size small is an ideal fit for women

Specifications

Certifications/Compliance 21 CFR, 170-199
Construction Seamless Knit
Coating Material Uncoated
Color Natural
Exterior Finish No Texture
Grade Economy
Hand Ambidextrous
Material Polycotton
Product Type Seamless Knit Gloves
Size Small
Style Reversible
For Use With (Application) Assembly, Food Processing, Parts Handling, and Warehousing
Glove Cuff Style Continuous Knit
Hem Color Yellow
Series 35-C103
Yarn Gauge 7
Length (English) 8.4 in.
Length (Metric) 21.5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.