Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ MaxiCut™ Dry Dyneema™ / Engineered Yarn Gloves, Nitrile Coating

Catalog No. 19152945 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
Glove Cuff Color:
Blue
Gray
Orange
Red
White
Yellow
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Glove Cuff Color
19-152-945 X-Large Yellow
19-152-941 X-Small Blue
19-152-942 Small Red
19-152-943 Medium Orange
19-152-944 Large White
19-152-946 2X-Large Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-152-945 Supplier PIP™ Supplier No. 19D475/XL64868
This item is not returnable. View return policy

Seamless Knit Dyneema™ / Engineered Yarn Gloves combines cut protection with comfort, flexibility and dexterity for dry environments.

  • Micro-foam nitrile coating is compatible with light oils and will provide a secure grip and abrasion resistance
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Unique high performance yarns and fibers offer cut protection while maintaining high levels of comfort
  • Reinforcement between the thumb and forefinger, improves cut resistance and extends glove life in an inherently weak area
  • Coating offers 360° breathability
  • Smooth and rounded fingertips improve sensitivity
  • Optimized grip delivered through micro-cup finish
  • All ingredients used in the manufacturing and construction of this product are compliant with the REACH legislation
  • Laundered prior to final packaging making it clean and safe for use

Specifications

ANSI Abrasion Rating ANSI 4
Certifications/Compliance ASTM F2992-15 ANSI Cut Level A4
Certifications/Compliance (Europe) CE
Coating Material Nitrile Foam
Color Gray
Cut Resistance Rating ANSI A4
Material Dyneema™, Nitrile, Lycra
Product Type Cut-resistant Gloves
Size X-Large
EN388 Standard EN 388:4443C
For Use With (Application) Electronics, glass cutting, metal handling and fabrication, food and meat processing
Glove Cuff Color Yellow
Glove Cuff Style Knit
Model 19-D475/XL
Series MaxiCut™ Dry
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.