Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Kut-Gard™ Seamless Knit Dyneema™ Medium Weight Glove
GSA_VA

Catalog No. 19100937 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
Glove Cuff Color:
Blue
Gray
Red
White
Yellow
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Glove Cuff Color
19-100-937 Small White
19-100-936 X-Small Blue
19-100-938 Medium Red
19-100-939 Large Yellow
19-100-940 X-Large Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-100-937 Supplier PIP™ Supplier No. 17D300/S
Add to Cart
Add to Cart
This item is not returnable. View return policy

Medium weight uncoated gloves feature seamless knit construction and provide comfort without sacrificing dexterity. Ideal for use in the food industry or as a glove liner.

  • Medium weight Dyneema™ shell provides dexterity, tactile sensitivity and cut resistance
  • Ideal for use as a glove liner
  • Ideal for food processing
  • Color coded cuff hems for easy size identification
  • Launderable for extended life and to reduce replacement costs
  • Complies with FDA food handling requirements 21 CFR, Part 177

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Certifications/Compliance Compliant with Federal regulations for food contact, 21CFR, 170-199
Construction Seamless Knit
Coating Material Uncoated
Color White
Cut Resistance Rating ANSI A2
Hand Specific
Latex Free Yes
Material Dyneema
Product Type Cut-resistant Gloves
Size Small
For Use With (Application) Food industry, use as a glove liner
Glove Cuff Color White
Glove Cuff Style Knit
Model 17-D300/S
Series Kut-Gard™ 17-D300
Yarn Gauge 7
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.