Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Polyurethane Coated Seamless Knit Polyester Gloves

Catalog No. 19046392 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
2X-Large
Large
Medium
Small
X-Large
X-Small
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-046-392 X-Small
19-046-393 Small
19-046-394 Medium
19-046-395 Large
19-046-396 X-Large
19-046-397 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-046-392 Supplier PIP™ Supplier No. 33G115/XS
Add to Cart
Add to Cart
This item is not returnable. View return policy

Seamless knit polyester shell offers increased comfort, finger dexterity, and breathability. Polyurethane coated flat grip on the palm and fingers provides tactility in dry and slightly oily conditions.

  • Seamless knit polyester shell increases comfort, finger dexterity, and breathability
  • Polyurethane coating takes on the surface properties of the glove liner to provide a tactile grip in dry and slightly oily conditions
  • Ideal for applications that require dexterity for precise handling
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Breathable back for comfort

Specifications

ANSI Abrasion Rating ANSI 2
Construction Seamless Knit
Coating Material Polyurethane
Color Gray
Exterior Finish No Texture
Hand Specific
Material Polyester
Product Type Seamless Knit Gloves
Size X-Small
Style Standard
For Use With (Application) Electronics Assembly, Inspection, and Quality Control
Glove Cuff Style Knit
Series 33-G115
Yarn Gauge 13
Length (English) 8.5 in.
Length (Metric) 21.5 cm
Show More Show Less

WARNING: Cancer - www.P65Warnings.ca.gov

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.