Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Seasonal H1N1 Hemagglutinin Antibody (7H12F6) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP241328
Description
Seasonal H1N1 Hemagglutinin Monoclonal specifically detects Seasonal H1N1 Hemagglutinin in Influenza A Virus H1N1, Virus samples. It is validated for Western Blot, ELISA.Specifications
Seasonal H1N1 Hemagglutinin | |
Monoclonal | |
1 mg/mL | |
Western Blot, ELISA 1 ug/mL | |
Hemagglutinin, Hemagglutinin HA1 chain, Hemagglutinin HA2 chain | |
Antibody was raised against a synthetic peptide from the Hemagglutinin protein. Amino Acid Squence: PNIGDQKALTHTEN | |
0.1 mg | |
Primary | |
Store at -20 degrees C. Avoid freeze/thaw cycles. |
Western Blot, ELISA | |
7H12F6 | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
Mouse | |
Affinity Purified | |
RUO | |
Influenza virus, Virus | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction