Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    Seasonal H1N1 Hemagglutinin Antibody (7H12F6), PerCP, Novus Biologicals™
 
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP241328PCP
Description
Seasonal H1N1 Hemagglutinin Monoclonal antibody specifically detects Seasonal H1N1 Hemagglutinin in Influenza virusN1, Virus samples. It is validated for Western Blot, ELISASpecifications
| Seasonal H1N1 Hemagglutinin | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Affinity purified | |
| RUO | |
| Influenza virus N1, Virus | |
| Purified | 
| Western Blot, ELISA | |
| 7H12F6 | |
| Western Blot, ELISA | |
| Hemagglutinin, Hemagglutinin HA1 chain, Hemagglutinin HA2 chain | |
| Antibody was raised against a synthetic peptide from the Hemagglutinin protein. Amino Acid Squence: PNIGDQKALTHTEN | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG1 | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
            