Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Seasonal H1N1 Hemagglutinin Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$196.10 - $499.50
Specifications
Antigen | Seasonal H1N1 Hemagglutinin |
---|---|
Concentration | 1 mg/mL |
Applications | ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Seasonal H1N1 Hemagglutinin Polyclonal specifically detects Seasonal H1N1 Hemagglutinin in Influenza A Virus H1N1, Virus samples. It is validated for ELISA.Specifications
Seasonal H1N1 Hemagglutinin | |
ELISA | |
Unconjugated | |
RUO | |
PBS with 0.02% Sodium Azide | |
Antibody was raised against a synthetic peptide from the Hemagglutinin protein. The peptide sequence is unique from the peptide sequence for product NBP2-41104 and NBP2-41108. This antibody is a cognate pair with antibody NBP2-41107. The immunogen is located within amino acids 180 - 230 of Seasonal H1N1 Hemagglutinin. Amino Acid Squence: PNIGDQKALTHTEN | |
Primary | |
This antibody is specific for the seasonal H1N1 influenza Hemagglutinin and will not recognize the corresponding Hemagglutinin sequence from the swine-origin H1N1 influenza (A/California/14/2009 (H1N1)). Will not cross-react with peptide corresponding to the swine-origin H1N1 influenza Hemagglutinin. |
1 mg/mL | |
Polyclonal | |
Rabbit | |
Influenza virus, Virus | |
Hemagglutinin, Hemagglutinin HA1 chain, Hemagglutinin HA2 chain | |
IgG | |
Affinity Purified | |
68 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title