Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Seasonal H1N1 Neuraminidase Antibody (10C5E11), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP241330B
Description
Seasonal H1N1 Neuraminidase Monoclonal specifically detects Seasonal H1N1 Neuraminidase in Influenza A Virus H1N1, Virus samples. It is validated for ELISA.Specifications
Seasonal H1N1 Neuraminidase | |
Monoclonal | |
1 mg/mL | |
ELISA | |
EC=3.2.1.18, Neuraminidase | |
Antibody was raised against a synthetic peptide from the seasonal influenza (H1N1) neuraminidase protein. | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG2b |
ELISA | |
10C5E11 | |
Biotin | |
PBS with 0.05% Sodium Azide | |
Mouse | |
Protein A purified | |
RUO | |
Influenza virus, Virus | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction