Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Seasonal H1N1 Neuraminidase Antibody (6F4C8) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$196.10 - $499.50
Specifications
Antigen | Seasonal H1N1 Neuraminidase |
---|---|
Concentration | 1 mg/mL |
Applications | ELISA |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Seasonal H1N1 Neuraminidase Monoclonal specifically detects Seasonal H1N1 Neuraminidase in Influenza A Virus H1N1, Virus samples. It is validated for ELISA.Specifications
Seasonal H1N1 Neuraminidase | |
ELISA | |
Unconjugated | |
RUO | |
PBS with 0.02% Sodium Azide | |
Antibody was raised against a synthetic peptide from the seasonal influenza (H1N1) neuraminidase protein. | |
Primary |
1 mg/mL | |
Monoclonal | |
Mouse | |
Influenza virus, Virus | |
EC=3.2.1.18, Neuraminidase | |
IgG2b | |
Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title