Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    Seasonal H1N1 Nucleocapsid Protein Antibody, mFluor Violet 500 SE, Novus Biologicals™
 
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP241114MFV500
Description
Seasonal H1N1 Nucleocapsid Protein Polyclonal antibody specifically detects Seasonal H1N1 Nucleocapsid Protein in Influenza virusN1, Virus samples. It is validated for ELISASpecifications
| Seasonal H1N1 Nucleocapsid Protein | |
| Polyclonal | |
| ELISA | |
| Nucleocapsid protein, Nucleoprotein, Protein N | |
| Antibody was raised against a synthetic peptide from the seasonal H1N1 NP protein. The immunogen is located within amino acids 350 - 400 of Seasonal H1N1 Nucleocapsid Protein. Amino Acid Squence: NMDNMGSS | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG | 
| ELISA | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Influenza virus N1, Virus | |
| Purified | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
            