Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hand2Mind Seasons and Weather Pattern Block Puzzle Set

Click to view available options
Grade:
K to 12
Description
- From dressing for the season to understanding weather patterns, this set helps children learn about the connection between sustainability, the seasons, and the weather.
- Children complete the 20 engaging activities and uncover information about appropriate seasonal activities, how to dress for the weather, the connections between weather and the environment, and so much more.
Specifications
Specifications
Product Type | Block Puzzle |
Grade | K to 12 |
Class Size | 1 Student |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction