Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Sebacoyl Chloride
SCI_ED

Catalog No. s25517 Shop All Fisherbrand Products
Click to view available options
Quantity:
25 mL
Packaging:
Solution
This item is not returnable. View return policy
This item is not returnable. View return policy

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 111-19-3, 7647-01-0
Molecular Formula C10H16Cl2O2
Molecular Weight (g/mol) 239.136
InChI Key WMPOZLHMGVKUEJ-UHFFFAOYSA-N
Synonym sebacoyl chloride, sebacyl chloride, sebacoyl dichloride, sebacic acid dichloride, decanedioyl chloride, sebacoylchloride, octane-1,8-dicarbonyl chloride, sebacoyl chloride, cp, acmc-1byrk, decanedioic acid chloride
PubChem CID 66072
IUPAC Name decanedioyl dichloride
SMILES C(CCCCC(=O)Cl)CCCC(=O)Cl

Specifications

Density 1.121g/mL
Assay Percent Range >92%
Packaging Solution
Linear Formula COCl(CH2)8COCl
Quantity 25 mL
Formula Weight 239.14
Chemical Name or Material Sebacoyl Chloride
ShelfLife 1 year
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.