Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

sec-Butyl Alcohol, 98.5%, Spectrum™ Chemical
SDP

Catalog No. 18613085 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 mL
1 L
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-613-085 20 L Steel Can
18-613-084 100 mL Amber Glass Bottle
18-613-083 1 L Amber Glass Bottle
18-613-087 4 L Amber Glass Bottle
18-613-086 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-613-085 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. B118720LTMT
Add to Cart
Add to Cart

Chemical Identifiers

CAS 78-83-1
Molecular Weight (g/mol) 74.12

Specifications

CAS Max % 1
Quantity 20 L
Packaging Steel Can
Boiling Point 94°C to 99.5°C
Percent Purity 0.985
Grade Ungraded
Chemical Name or Material Sec-Butyl Alcohol

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.