Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

sec-Butylbenzene, 99%

Catalog No. p-7026761
Click to view available options
Quantity:
25 mL

CAS: 135-98-8 | C10H14 | 134.222 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135-98-8
Molecular Formula C10H14
Molecular Weight (g/mol) 134.222
MDL Number MFCD00009329
InChI Key ZJMWRROPUADPEA-UHFFFAOYSA-N
Synonym sec-butylbenzene, 2-phenylbutane, 1-methylpropyl benzene, secondary butylbenzene, benzene, 1-methylpropyl, s-butylbenzene, benzene, sec-butyl, 1-sec-butylbenzene, methylpropyl benzene, secbutylbenzene
PubChem CID 8680
ChEBI CHEBI:35097
IUPAC Name butan-2-ylbenzene
SMILES CCC(C)C1=CC=CC=C1

Specifications

Melting Point -75°C
Density 0.862
Boiling Point 173°C to 174°C
Flash Point 52°C (125°F)
Odor Aromatic
Refractive Index 1.49
Quantity 25 mL
UN Number UN2709
Beilstein 1903902
Merck Index 14,1550
Formula Weight 134.22
Percent Purity 99%
Chemical Name or Material sec-Butylbenzene
Show More Show Less
Hazard Category H226-H315-H319
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: BUTYL BENZENES
EINECSNumber 205-227-0
RTECSNumber CY9100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.