Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SEC11A Monoclonal Antibody (OTI1A12)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA527472

Catalog No. PIMA527472


Only null left
Add to Cart

Description

Description

SEC11A Monoclonal Antibody for Western Blot, IHC (P)

This gene encodes a member of the peptidase S26B family. The encoded protein is an 18kDa subunit of the signal peptidase complex and has been linked to cell migration and invasion, gastric cancer and lymph node metastasis. Alternative splicing results in multiple transcript variants. A related pseudogene has been identified on chromosome 8.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SEC11A
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P67812
SEC11A
Full length human recombinant protein of SEC11A produced in E.coli.
100 μL
Primary
Human
Antibody
IgG2b
Immunohistochemistry (Paraffin), Western Blot
OTI1A12
Unconjugated
SEC11A
1810012E07Rik; 18kDa; Endopeptidase SP18; microsomal signal peptidase 18 kDa subunit; SEC11 homolog A; SEC11 homolog A (S. cerevisiae); SEC11 homolog A, signal peptidase complex subunit; SEC11A; Sec11l1; Sec11-like 1; SEC11-like protein 1; sid 2895; Sid2895; Sid2895p; signal peptidase complex (18kD); signal peptidase complex 18; signal peptidase complex 18kD; signal peptidase complex catalytic subunit SEC11A; SPase 18 kDa subunit; SPC18; SPCS4A
Mouse
Affinity Chromatography
RUO
23478
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.