Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SEC11A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA549273
Description
Predicted to react with mouse, rat and bovine based on sequence homology.
This gene encodes a member of the peptidase S26B family. The encoded protein is an 18kDa subunit of the signal peptidase complex and has been linked to cell migration and invasion, gastric cancer and lymph node metastasis. Alternative splicing results in multiple transcript variants. A related pseudogene has been identified on chromosome 8.
Specifications
SEC11A | |
Polyclonal | |
Unconjugated | |
SEC11A | |
1810012E07Rik; 18kDa; Endopeptidase SP18; microsomal signal peptidase 18 kDa subunit; SEC11 homolog A; SEC11 homolog A (S. cerevisiae); SEC11 homolog A, signal peptidase complex subunit; SEC11A; Sec11l1; Sec11-like 1; SEC11-like protein 1; sid 2895; Sid2895; Sid2895p; signal peptidase complex (18kD); signal peptidase complex 18; signal peptidase complex 18kD; signal peptidase complex catalytic subunit SEC11A; SPase 18 kDa subunit; SPC18; SPCS4A | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
23478 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P67812 | |
SEC11A | |
KLH conjugated synthetic peptide between 95-123 amino acids from the Central region of human SC11A. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction