Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SEC11A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120093
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120093 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120093 Supplier Invitrogen™ Supplier No. PA5120093
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: SGC-7901, HT-29, Raji, HepG2, A-549, Mouse kidney, Mouse liver, Mouse lung, Rat brain. The target is usually found in the following locations: Endoplasmic reticulum membrane, Microsome membrane, Single-pass type II membrane protein. Immunogen sequence: KGLMVITGSE SPIVVVLSGS MEPAFHRGDL LFLTNRVEDP IRVGEIVVFR IEGREIPIVH RVLKIHEKQN GHIKFLTKGD NNAVDDRGLY KQGQHWLEKK DVVGRARGF.

This gene encodes a member of the peptidase S26B family. The encoded protein is an 18kDa subunit of the signal peptidase complex and has been linked to cell migration and invasion, gastric cancer and lymph node metastasis. Alternative splicing results in multiple transcript variants. A related pseudogene has been identified on chromosome 8.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEC11A
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.08 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene SEC11A
Gene Accession No. P42667, P67812, Q9R0P6
Gene Alias 1810012E07Rik; 18kDa; Endopeptidase SP18; microsomal signal peptidase 18 kDa subunit; SEC11 homolog A; SEC11 homolog A (S. cerevisiae); SEC11 homolog A, signal peptidase complex subunit; SEC11A; Sec11l1; Sec11-like 1; SEC11-like protein 1; sid 2895; Sid2895; Sid2895p; signal peptidase complex (18kD); signal peptidase complex 18; signal peptidase complex 18kD; signal peptidase complex catalytic subunit SEC11A; SPase 18 kDa subunit; SPC18; SPCS4A
Gene Symbols SEC11A
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 37-145 of human SEC11A (NP_055115.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23478, 56529, 65166
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.