Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SEC11C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555427
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555427 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555427 Supplier Invitrogen™ Supplier No. PA555427
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MVRAGAVGAH LPASGLDIFG DLKKMNKRQL YYQV Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 88%.

Component of the microsomal signal peptidase complex which removes signal peptides from nascent proteins as they are translocated into the lumen of the endoplasmic reticulum.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEC11C
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SEC11C
Gene Accession No. Q9BY50
Gene Alias 1810029G24Rik; microsomal signal peptidase 21 kDa subunit; SEC11 homolog C; SEC11 homolog C (S. cerevisiae); SEC11 homolog C, signal peptidase complex subunit; Sec11c; Sec11l3; Sec11-like 3; SEC11-like protein 3; signal peptidase complex 21; signal peptidase complex catalytic subunit SEC11C; SPase 21 kDa subunit; SPC21; SPCS4C
Gene Symbols SEC11C
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SEC11C. Recombinant protein control fragment (Product #RP-88702).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90701
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.